Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho-HDAC1 (Ser406) Mouse anti-Human,Bovine,Monkey,Rat,Rabbit, KHL, Clone: BT-15, MilliporeSigma™

Catalog No. MABE18211MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE18211MI 100 μg
MABE18212MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE18211MI Supplier MilliporeSigma Supplier No. MABE1821100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

phospho-HDAC1 (Ser406) Monoclonal antibody specifically detects phospho-HDAC1 (Ser406) in Human,Bovine,Monkey,Rat,Rabbit samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-HDAC1 (Ser406)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BT-15
Conjugate KLH
Gene Accession No. NP_004955
Gene Alias Histone deacetylase 1;HD1
Gene Symbols HDAC1, RPD3L1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 19 amino acids surrounding phosphoserine 406 from the C-terminal region of human Histone deacetylase 1.
Molecular Weight of Antigen 60 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity Clone BT-15 is a mouse monoclonal antibody that specifically detects Histone deacetylase phosphorylated on serine 406 in mitotic synchronized cells. It targets an epitope within 19 amino acids from the C-terminal region.
Target Species Human, Bovine, Monkey, Rat, Rabbit
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.