Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HDAC1 (Ser421) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536810
Description
This antibody detects endogenous protein at a molecular weight of 55 kDa. Purity is >95% by SDS-PAGE.
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
Specifications
| Phospho-HDAC1 (Ser421) | |
| Polyclonal | |
| Unconjugated | |
| HDAC1 | |
| DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 297893, 3065, 433759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O09106, Q13547, Q4QQW4 | |
| HDAC1 | |
| Synthetic phosphopeptide derived from human HDAC1 around the phosphorylation site of Serine 421. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction