Learn More
Invitrogen™ Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) Monoclonal Antibody (B.544.8)

Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) |
| Applications | ChIP Assay, Western Blot |
| Classification | Monoclonal |
| Clone | B.544.8 |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 |
| Gene | HDAC4 |
| Gene Accession No. | P56524, Q6NZM9, Q8C2B3, Q8WUI4, Q9UQL6, Q9Z2V6 |
| Gene Alias | 4932408F19Rik; 5830434K02Rik; AHO3; AI047285; AI426555; Antigen NY-CO-9; BDMR; HA6116; HD4; HD5; HD7; HD7a; Hdac4; HDAC-4; HDAC5; Hdac7; hdac-7; HDAC7A; HDACA; HDAC-A; Histone deacetylase 4; histone deacetylase 5; Histone deacetylase 7; histone deacetylase 7A; histone deacetylase A; histone deacetylase mHDA1; KIAA0288; KIAA0600; mFLJ00062; mHDA1; mKIAA0600; NY-CO-9 |
| Gene Symbols | HDAC4, HDAC5, HDAC7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.