Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537835
Description
A suggested positive control for Western blot is 293 cells; suggested positive control for IHC is human breast carcinoma.
Histone deacetylase (HDAC) and histone acetyltransferase (HAT) are enzymes that regulate transcription by selectively deacetylating or acetylating the eta-amino groups of lysines located near the amino termini of core histone proteins (1). Eight members of HDAC family have been identified in the past several years (2,3). These HDAC family members are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8, each of which contains a deacetylase domain exhibiting from 45 to 93% identity in amino acid sequence. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about twofold larger than those of the class I members, and the deacetylase domains are present within the C-terminal regions, except that HDAC-6 contains two copies of the domain, one within each of the N-terminal and C-terminal regions. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns (3). These results suggested that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.
Specifications
Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220) | |
Polyclonal | |
Unconjugated | |
HDAC4 | |
AHO3; Antigen NY-CO-9; BDMR; DKFZp779K1053; HA6116; HD4; HD5; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC5; HDAC7; HDAC7B; HDAC9; hdac-9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; histone deacetylase 4; histone deacetylase 4/5-related protein; histone deacetylase 5; Histone deacetylase 7B; histone deacetylase 9; histone deacetylase A; Histone deacetylase-related protein; KIAA0288; KIAA0600; KIAA0744; MEF-2 interacting transcription repressor (MITR) protein; MEF2-interacting transcription repressor MITR; MITR; NY-CO-9 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10014, 9734, 9759 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P56524, Q9UKV0, Q9UQL6 | |
HDAC4, HDAC5, HDAC9 | |
Peptide sequence around phosphorylation site of serine 246/259/220 (T-A-S(p)-EP) derived from Human HDAC4/HDAC5/HDAC9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction