Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HDAC4 (Ser632) Recombinant Rabbit Monoclonal Antibody (0A1R4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551517
Description
Immunogen sequence: AQSSP.
Histone deacetylase 4 (HDAC4) is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3, and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. HDAC4 is also involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D. It is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer. Mutations to the HDAC4 gene have been found in Brachydactyly-mental retardation syndrome (BDMR).
Specifications
Phospho-HDAC4 (Ser632) | |
Recombinant Monoclonal | |
0.071 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P56524 | |
HDAC4 | |
A synthetic phosphorylated peptide around S632 of human Phospho-HDAC4-S632 (NP_006028.2). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Western Blot | |
0A1R4 | |
Unconjugated | |
HDAC4 | |
4932408F19Rik; AHO3; AI047285; BDMR; DKFZp779K1053; HA6116; HD4; HD7; HD7b; HD9; HDAC; HDAC4; HDAC-4; HDAC7; HDAC7B; HDAC9; HDAC9B; HDAC9FL; HDACA; HDAC-A; HDRP; Histone deacetylase 4; histone deacetylase A; KIAA0288; KIAA0744; MITR | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9759 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction