Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Phospho-HDAC5 (Ser259) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538116
Description
HDAC5 is a member of the class II mammalian histone deacetylase family, which is structurally related to yeast HDA1. Human HDAC5 is composed of 1122 amino acid residues. The deacetylase domain of HDAC5 is located at the C-terminal half of the molecule. The N-terminal non-deacetylase domain does not show any significant homology with any published sequence.Both domains are required for HDAC5-mediated repression of gene transcription. HDAC5 interacts with a growing number of transcriptional factors including MEF2A as well as other HDAC proteins. The interacting complexes bind to specific regions of chromatin and regulate gene transcription in these regions. 
            Specifications
| Phospho-HDAC5 (Ser259) | |
| Polyclonal | |
| Unconjugated | |
| HDAC5 | |
| Histone deacetylase 5, Short name=HD5, Antigen NY-CO-9, HDAC-5, KIAA0600 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10014, 15184 | |
| -20°C | |
| Liquid | 
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UQL6, Q9Z2V6 | |
| HDAC5 | |
| A synthetic phosphopeptide derived from human HDAC5 around the phosphorylation site of Ser259 (T-A-SP-E-P) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                             
                             
                             
                            