Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HDAC7 (Ser486) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106090
Description
Antibody detects endogenous levels of HDAC7 only when phosphorylated at Ser486.
Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation/deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Phospho-HDAC7 (Ser486) | |
| Polyclonal | |
| Unconjugated | |
| HDAC7 | |
| 5830434K02Rik; HD7; HD7a; hdac4l; Hdac7; hdac-7; Hdac7a; histone deacetylase 7; Histone deacetylase 7A; mFLJ00062 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 51564, 56233 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C2B3, Q8WUI4 | |
| HDAC7 | |
| A synthesized peptide derived from human HDAC7(Accession Q8WUI4), corresponding to amino acid residues around phosphorylated Ser486. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction