Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HIC5 (Tyr60) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105804 Supplier Invitrogen™ Supplier No. PA5105804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Hic-5 only when phosphorylated at Tyr60.

This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HIC5 (Tyr60)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Tgfb1i1
Gene Accession No. O43294, Q62219, Q99PD6
Gene Alias Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; androgen receptor-associated protein of 55 kDa; Ara55; Hic5; hic-5; hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Tgfb1i1; transforming growth factor beta 1 induced transcript 1; transforming growth factor beta-1-induced transcript 1 protein; TSC-5
Gene Symbols Tgfb1i1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TGFB1I1(Accession O43294), corresponding to amino acid residues around phosphorylated Tyr60.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21804, 7041, 84574
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.