Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-HICE1 pS70 (Ser70) Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992023
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2023 100 μg
50-199-2024 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2023 Supplier Rockland Immunochemicals Supplier No. 600401C52
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Hice1 pS70 affinity purified antibody is directed against the phosphorylated form of human Hice1 protein at the S70 residue. The antibody is specific for the phosphorylated form of Hice1. Reactivity with non-phosphorylated human Hice1 is minimal by ELISA and western blot. A BLAST analysis was used to suggest cross reactivity with Hice1 from human based on 100% sequence homology with the immunogen. Reactivity against homologues from other sources is not known.

Hice1 contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.

Specifications

Antigen Phospho-HICE1 pS70 (Ser70)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene HAUS8
Gene Accession No. Q9BT25
Gene Alias 2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; Haus8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; HICE1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; Sarcoma antigen NY-SAR-48 homolog
Gene Symbols HAUS8
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93323
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.