Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HICE1 (Ser19, Ser20) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534977 Supplier Invitrogen™ Supplier No. PA534977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, K562, Blocking peptide (GTX70011-PEP). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Hice1 contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HICE1 (Ser19, Ser20)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HAUS8
Gene Accession No. Q9BT25
Gene Alias 2410004L22Rik; 4HAUS augmin-like complex, subunit 8; DGT4; HAUS augmin like complex subunit 8; HAUS augmin-like complex subunit 8; HAUS augmin-like complex, subunit 8; HAUS8; HEC1/NDC80 interacting, centrosome associated 1; HEC1/NDC80-interacting centrosome-associated protein 1; Hec1-interacting and centrosome-associated 1; Hice1; MGC20533; NY SAR 48; NYSAR48; NY-SAR-48; Sarcoma antigen NY-SAR-48; sarcoma antigen NY-SAR-48 homolog
Gene Symbols HAUS8
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to the amino acids around Ser 19/20 conjugated to KLH.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93323
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.