Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HIPK2 (Tyr361) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513045
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
HIPK2 is a nuclear serine/threonine kinase that is linked to transcriptional regulation and apoptosis.
Specifications
| Phospho-HIPK2 (Tyr361) | |
| Polyclonal | |
| Unconjugated | |
| HIPK2 | |
| 1110014O20Rik; B230339E18Rik; cb1009; hHIPk2; HIPK2; homeodomain interacting protein kinase 2; homeodomain-interacting protein kinase 2; Nbak1; Nuclear body-associated kinase 1; PRO0593; si:dkey-16j24.2; si:dkey-239k7.1; Sialophorin tail-associated nuclear serine/threonine-protein kinase; Stank; wu:fk56d12 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 28996 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Dot Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H2X6 | |
| HIPK2 | |
| KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y361 of human HIPK2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction