Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H1.3 (Thr18) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5101017
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101017 Supplier Thermo Scientific Supplier No. PA5101017

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Histone H1.3 only when phosphorylated at Thr18.

Histones are basic nuclear proteins responsible for nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H1 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Histone H1.3 (Thr18)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H1-3
Gene Accession No. P16402, P43277
Gene Alias H1 histone family, member 3, H1 VAR.4, H1.3, H1D, H1f3, H1s-2, H1s-4, Hist1h1d, histone 1, family 3, histone 1, H1d, histone cluster 1, H1d, histone H1.3, Histone H1c, histone H1s-2
Gene Symbols H1-3, HIST1H1D
Host Species Rabbit
Immunogen A synthesized peptide derived from human HIST1H1E(Accession P10412), corresponding to amino acid residues around phosphorylated Thr18.
Purification Method sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14957, 3007
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.