Learn More
Invitrogen™ Phospho-Histone H1 (Thr3) Polyclonal Antibody

Description
This antibody is predicted to react with mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-Histone H1 (Thr3) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Whole serum with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | H1-5 |
| Gene Accession No. | D3ZBN0, D4A3K5, P07305, P10922, P15864, P16401, P16403, P43275, P43276, P43278, Q02539 |
| Gene Alias | 0610008C09Rik; CTL-1; H1; H1 antibody; H1 histone family member 0; H1 histone family, member 0; H1 histone family, member 1; H1 histone family, member 2; H1 histone family, member 5; H1 VAR.1; H1 VAR.3; H1 VAR.5; H1(0); H1.0; H1.0, H1(0), H1-0; H1.1; H1.12 linker histone, cluster member; H1.2; H1.2 linker histone, cluster member; H1.5; H1.9; H10; H1-0; H1-1; H1-12; H1-2; H1A; H1B; H1c; H1F0; H1f1; H1F2; H1F3; H1F5; H1FV; H1s-1; H1s-3; H1var1; H1var3; Hils1; His1a; HIST1; HIST1H1A; HIST1H1B; HIST1H1B antibody; HIST1H1C; HIST1H1D; HIST1H1E; Histone 1 antibody; histone 1, family 5; histone 1, H1a; histone 1, H1b; histone 1, H1c; histone cluster 1, H1a; histone cluster 1, H1b; histone cluster 1, H1c; histone cluster 1, H1e; histone H1; histone H1'; Histone H1(0); histone H1.0; Histone H1.0, N-terminally processed; Histone H1.1; histone H1.2; Histone H1.3; Histone H1.5; histone H1a; Histone H1b; histone H1c; Histone H1d; histone H1-like protein in spermatids 1; histone H1s-1; Histone H1s-2; Histone H1s-3; histone linker H1 domain, spermatid-specific 1 (pseudogene); spermatid-specific linker histone H1-like protein; TISP64 |
| Gene Symbols | H1-0, H1-1, H1-2, H1-5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.