Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-Histone H2A (Thr121) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104780 Supplier Invitrogen™ Supplier No. PA5104780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of Histone H2A only when phosphorylated at Threonine 121.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Histone H2A (Thr121)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene H2AZ1
Gene Accession No. C0HKE1, C0HKE3, C0HKE4, C0HKE5, C0HKE6, P04908, P0C0S5, P0C0S6, P0C0S7, P0C0S8, P16104, P20671, P27661, Q00728, Q00729, Q8CGP6, Q96A08, Q96KK5, Q96QV6, Q99878
Gene Alias 1190022L06; Acetyl-Histone H2; AW228881; dJ160A22.4; dJ86C11.1; gammaH2ax; H2A; H2A 615 antibody; H2A GL101 antibody; H2A histone; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member V; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1 antibody; H2A.1b; H2A.2; H2A.3; H2a.x; H2A.Z; H2A.Z1; H2a.z-1; H2A.Zl; h2a.zl1; h2a.zl2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/q; H2A/S; H2A/X; H2A/z; H2a-291A; H2a-53; h2a-A; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; h2ad; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2AFQ; H2AFV; H2afx; H2afz; h2afz.L; H2AG; H2A-GL101; H2AH; h2ai; H2A-III; H2ak; H2AK5ac; H2AX; H2AX histone; H2Az; H2AZ histone; HIS; HIST1H2A3; Hist1h2aa; Hist1h2ab; Hist1h2ac; Hist1h2ad; HIST1H2AE; HIST1H2AE antibody; HIST1H2AG; Hist1h2ah; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1h2an; Hist1h2ao; Hist1h2ap; Hist1h2ba; hist2h2ab; hist2h2ab.S; HIST2H2AC; HIST3H2A; Hist5-2ax; histone 1, H2ab; histone 1, H2ad; histone 1, H2ae; histone 1, H2ag; histone 1, H2ah; histone 1, H2ai; histone 1, H2aj; histone 1, H2ba; histone 2, H2ac; histone 2, H2ai; histone 5 protein 2ax; histone cluster 1, H2A, III (similar to human histone cluster 2, class H2A, member C); histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2aj; histone cluster 1, H2ba; histone cluster 2, H2ab S homeolog; histone cluster 2, H2ac; histone H2A; histone H2A type 1; Histone H2A type 1-B; Histone H2A type 1-B/E; Histone H2A type 1-C; Histone H2A type 1-D; Histone H2A type 1-E; Histone H2A type 1-G; Histone H2A type 1-H; Histone H2A type 1-I; Histone H2A type 1-J; Histone H2A type 1-N; Histone H2A type 1-O; Histone H2A type 1-P; Histone H2A type 2-C; histone H2A type 4; Histone H2A, testis; Histone H2A.2; Histone H2A.3; histone H2A.x; Histone H2A.Z; histone H2A.Zl2; histone H2A.Z-like; histone H2A/a; Histone H2A/e; Histone H2A/g; histone H2A/m; histone H2A/p; Histone H2A/ptl; Histone H2A/q; histone H2A/s; histone H2AD; histone H2AE; histone H2A-GL101; histone H2A-III; histone H2A-IV; histone H2AX; Histone H2B type 1-A; histone H2B, testis; histone IIa; hypothetical protein; LOC680322; MGC74460 antibody; pH2A/f; RCJMB04_2h11; RGD1562827; RGD1566119; similar to Histone H2A type 1; testis-specific histone 2a; testis-specific histone 2b; testis-specific histone H2B; TH2A; Th2b; XELAEV_18004421mg; XELAEV_18005732mg; XH2AX; XH2AZ
Gene Symbols H2ac1, H2AC11, H2AC12, H2AC13, H2AC14, H2AC4, H2AC7, H2AC8, H2AFX, H2AX, H2AZ1, H2bc1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HIST1H2AG; HIST1H2AI; HIST1H2AK; HIST1H2AL; HIST1H2AM(Accession P0C0S8), corresponding to amino acid residues around phosphorylated Thr121.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15270, 221613, 24828, 24829, 255626, 3012, 3013, 3014, 3015, 319165, 319166, 319167, 319168, 319172, 319174, 319191, 500987, 51788, 58940, 680322, 8329, 8331, 8335, 85235, 8969
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.