Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-Histone H2A (Tyr142) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144298
Description
Phospho-Histone H2A (Tyr142) Polyclonal Antibody for ICC/IF, ELISA
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
Phospho-Histone H2A (Tyr142) | |
Polyclonal | |
Unconjugated | |
H2AZ1 | |
1190022L06; Acetyl-Histone H2; AW228881; dJ160A22.4; dJ86C11.1; gammaH2ax; H2A; H2A 615 antibody; H2A GL101 antibody; H2A histone; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member V; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1 antibody; H2A.1b; H2A.2; H2A.3; H2a.x; H2A.Z; H2A.Z1; H2a.z-1; H2A.Zl; h2a.zl1; h2a.zl2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/q; H2A/S; H2A/X; H2A/z; H2a-291A; H2a-53; h2a-A; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; h2ad; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2AFQ; H2AFV; H2afx; H2afz; h2afz.L; H2AG; H2A-GL101; H2AH; h2ai; H2A-III; H2ak; H2AK5ac; H2AX; H2AX histone; H2Az; H2AZ histone; HIS; HIST1H2A3; Hist1h2aa; Hist1h2ab; Hist1h2ac; Hist1h2ad; HIST1H2AE; HIST1H2AE antibody; HIST1H2AG; Hist1h2ah; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; Hist1h2an; Hist1h2ao; Hist1h2ap; Hist1h2ba; hist2h2ab; hist2h2ab.S; HIST2H2AC; HIST3H2A; Hist5-2ax; histone 1, H2ab; histone 1, H2ad; histone 1, H2ae; histone 1, H2ag; histone 1, H2ah; histone 1, H2ai; histone 1, H2aj; histone 1, H2ba; histone 2, H2ac; histone 2, H2ai; histone 5 protein 2ax; histone cluster 1, H2A, III (similar to human histone cluster 2, class H2A, member C); histone cluster 1, H2aa; histone cluster 1, H2ab; histone cluster 1, H2ad; histone cluster 1, H2ae; histone cluster 1, H2ag; histone cluster 1, H2ah; histone cluster 1, H2ai; histone cluster 1, H2aj; histone cluster 1, H2ba; histone cluster 2, H2ab S homeolog; histone cluster 2, H2ac; histone H2A; histone H2A type 1; Histone H2A type 1-B; Histone H2A type 1-B/E; Histone H2A type 1-C; Histone H2A type 1-D; Histone H2A type 1-E; Histone H2A type 1-G; Histone H2A type 1-H; Histone H2A type 1-I; Histone H2A type 1-J; Histone H2A type 1-N; Histone H2A type 1-O; Histone H2A type 1-P; Histone H2A type 2-C; histone H2A type 4; Histone H2A, testis; Histone H2A.2; Histone H2A.3; histone H2A.x; Histone H2A.Z; histone H2A.Zl2; histone H2A.Z-like; histone H2A/a; Histone H2A/e; Histone H2A/g; histone H2A/m; histone H2A/p; Histone H2A/ptl; Histone H2A/q; histone H2A/s; histone H2AD; histone H2AE; histone H2A-GL101; histone H2A-III; histone H2A-IV; histone H2AX; Histone H2B type 1-A; histone H2B, testis; histone IIa; hypothetical protein; LOC680322; MGC74460 antibody; pH2A/f; RCJMB04_2h11; RGD1562827; RGD1566119; similar to Histone H2A type 1; testis-specific histone 2a; testis-specific histone 2b; testis-specific histone H2B; TH2A; Th2b; XELAEV_18004421mg; XELAEV_18005732mg; XH2AX; XH2AZ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221613, 255626, 3012, 3013, 3014, 3015, 8329, 8331, 8335, 85235, 8969 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P04908, P0C0S5, P0C0S8, P16104, P20671, Q96A08, Q96KK5, Q96QV6, Q99878 | |
H2ac1, H2AC11, H2AC12, H2AC13, H2AC14, H2AC4, H2AC7, H2AC8, H2AFX, H2AX, H2AZ1, H2bc1 | |
Peptide sequence around site of Phospho-Tyr (142) derived from Human Histone H2AX. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction