Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Histone H2A.X (Ser139) Polyclonal Antibody
Phospho-Histone H2A.X (Ser139) Polyclonal Antibody
Supplier: Thermo Scientific PA5117754
Description
Phospho-Histone H2A.X (Ser139) Polyclonal Antibody for Western Blot, ICC/IF, ELISA, DB

Specifications
Phospho-Histone H2A.X (Ser139) | |
Polyclonal | |
Unconjugated | |
H2AX | |
AW228881; gamma H2AX; gammaH2ax; H2A histone family member X; H2A histone family, member X; H2A.X; H2A.X variant histone; H2A/X; h2afx; H2ax; H2AX histone; Hist5-2ax; histone 5 protein 2ax; histone H2A.x; Histone H2a/x; Histone H2AX; RGD1566119; similar to H2A histone family, member X; zgc:56329 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3014 | |
-20°C or -80°C if preferred | |
Liquid |
Dot Blot, ELISA, Western Blot, Immunocytochemistry | |
2.75 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide | |
P16104 | |
H2AX | |
Phospho-Histone H2A.X (Ser139) synthetic peptide. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction