Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospho Histone H4 (Tyr51), Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABE290925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE290925UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE290925UL Supplier MilliporeSigma Supplier No. ABE290925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho Histone H4 (Tyr51) Antibody, Cat. No. ABE2909, is a highly specific rabbit polyclonal antibody that targets Histone H4 phosphorylated on Tyr51 and has been tested for use in in Immunocytochemistry, Immunofluorescence, and Western Blotting.

Histone H4 (UniProt: P62805 ) is encoded by the HIST14A (also known as H4/A, H4FA, HIST1H4B, H4/I, H4FI, HIST1H4C, H4/G, H4FG, HIST1H4D, H4/B, H4FB, HIST1H4E, H4/J, H4FJ, HIST1H4F, H4/C, H4FC, HIST1H4H, H4/H, H4FH, HIST1H4I, H4/M, H4FM, HIST1H4J, H4/E, H4FE, HIST1H4K, H4/D, H4FD, HIST1H4L, H4/K, H4FK, HIST2H4A, H4/N, H4F2, H4FN, HIST2H4, HIST2H4B, H4/O, H4FO, HIST4H4) gene (Gene ID: 121504, 554313, 8294, 8359, 8360, 8361, 8362, 8363, 8364, 8365, 8366, 8367, 8368, 8370) in human. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H4 is one of the main histone proteins that contribute to the chromatin structure in both prokaryotic and eukaryotic cells. Histone H4 can undergo several different types of epigenetic modifications that influence cellular processes, including methylation, acetylation, and phosphorylation. Acetylation at Lys6, Lys9, Lys13 and Lys17 is shown to occur in coding regions of the genome but not in heterochromatin. Monomethylation and asymmetric dimethylation at Arg4 is reported to favor acetylation at Lys9 and Lys13. Histone H4 can be phosphorylated by Tie2 on Tyr51, which is shown to preferentially occur in the presence of core histones. The phosphorylated H4 (H4Y51) is recognized by the proto-oncogene ABL1. Histone H4 is also phosphorylated on Ser48 by PAK2, which is shown to increase the association of H3.3-H4 with the histone chaperone HIRA, thus promoting nucleosome assembly of H3.3-H4 and inhibiting nucleosome assembly of H3.1-H4. Chromosomal aberrations involving histone H4 causes of B-cell non-Hodgkin lymphomas. (Ref.: Hossain, MB., et al. (2016). Sci. Adv. 2; e1501290).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Histone H4 (Tyr51)
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Tyr51 phosphorylation of Histone H4 in the presence of active recombinant Tie2, following analysis by Western blotting.  (Hossain, MB., et al. (2016). Sci. Adv. 2; e1501290). Immunofluorescence Analysis: A representative lot detected phospho Histone H4 (Tyr51) in HEK293 Tie2 cells treated with  Angiopoietin 1 ( ANG1) and ionizing radiation (Hossain, MB., et al. (2016). Sci. Adv. 2; e1501290).  
Formulation Purified rabbit polyclonal antibody containing PBS without preservatives and 40% glycerol.
Gene Alias H4;H4Y51;H4pY51
Gene Symbols HIST14A;H4/A;H4FA;HIST1H4B;H4/I;H4FI;HIST1H4C;H4/G;H4FG;HIST1H4D;H4/B;H4FB;HIST1H4E;H4/J;H4FJ;HIST1H4F;H4/C;H4FC;HIST1H4H;H4/H;H4FH;HIST1H4I;H4/M;H4FM;HIST1H4J;H4/E;H4FE;HIST1H4K;H4/D;H4FD;HIST1H4L;H4/K;H4FK;HIST2H4A;H4/N;H4F2;H4FN;HIST2H4;HIST2H4B;H4/O;H4FO;HIST4H4
Host Species Rabbit
Immunogen Two linear peptides (phosphorylated on Tyr 51 and non-phosphorylated) corresponding to 11 amino acids from the internal region of human Histone H4.
Molecular Weight of Antigen 11.38 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Histone H4 phosphorylated on Tyrosine 51. It targets an epitope within 11 amino acids surrounding phosphotyrosine 51.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.