Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-HNF4 (Ser313) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985823
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5823 Supplier Bioss Supplier No. BS4001R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.

Specifications

Antigen Phospho-HNF4 (Ser313)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HNF4A
Gene Accession No. P41235
Gene Alias alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; Hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; Nr2a1; NR2A21; Nuclear receptor 2A1; Nuclear receptor subfamily 2 group A member 1; TCF; Tcf14; TCF-14; Transcription factor 14; Transcription factor HNF-4
Gene Symbols HNF4A
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human HNF4 alpha around the phosphorylation site of Ser313.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3172
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.