Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HP1 gamma (Ser83) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110111
Description
Immunogen sequence: RKSLS.
At the nuclear envelope, the nuclear lamina and heterochromatin are adjacent to the inner nuclear membrane. The protein encoded by this gene binds DNA and is a component of heterochromatin. This protein also can bind lamin B receptor, an integral membrane protein found in the inner nuclear membrane. The dual binding functions of the encoded protein may explain the association of heterochromatin with the inner nuclear membrane.
Specifications
Phospho-HP1 gamma (Ser83) | |
Polyclonal | |
Unconjugated | |
Cbx3 | |
CBX3; chromobox 3; chromobox homolog 3; chromobox homolog 3 (HP1 gamma homolog, Drosophila); chromobox protein homolog 3; chromobox-like protein 3; HECH; heterochromatin protein 1 gamma; heterochromatin protein 1 homolog gamma; heterochromatin protein HP1 gamma; heterochromatin-like protein 1; HGNC:1553; HP1 gamma; HP1 gamma homolog; HP1g; HP1-GAMMA; HP1Hs-gamma; HPy; M32; Modifier 2 protein; OTTHUMP00000202463 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11335, 12417 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P23198, Q13185 | |
Cbx3 | |
A synthetic phosphorylated peptide around S83 of human HP1 gamma/CBX3 (NP_0092072). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction