Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HPK1 (Ser171) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512881
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512881 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512881 Supplier Invitrogen™ Supplier No. PA512881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology.

MAP4K1 (also known as HPK1) or Hematopoietic progenitor kinase 1 is a hematopoietic cell-restricted member of the Ste20 serine/threonine kinase super family. MAP4K1 is a tissue-specific upstream activator of the MEKK/JNK/SAPK signaling pathway. MAP4K1 diminishes T cell receptor (TCR) signaling activity and T cell proliferation by phosphorylating the adaptor protein SLP-76. The c-Jun amino-terminal kinases (JNKs)/stress-activated protein kinases (SAPKs) play a crucial role in stress responses in mammalian cells. The mechanism underlying this pathway in the hematopoietic system is unclear, but it is a key in understanding the molecular basis of blood cell differentiation. We have cloned a novel protein kinase, termed hematopoietic progenitor kinase 1 (HPK1), that is expressed predominantly in hematopoietic cells, including early progenitor cells. HPK1 is related distantly to the p21(Cdc42/Rac1)-activated kinase (PAK) and yeast STE20 implicated in the mitogen-activated protein kinase (MAPK) cascade. Expression of HPK1 activates JNK1 specifically, and it elevates strongly AP-1-mediated transcriptional activity in vivo. HPK1 binds and phosphorylates MEKK1 directly, whereas JNK1 activation by HPK1 is inhibited by a dominant-negative MEKK1 or MKK4/SEK mutant. Interestingly, unlike PAK65, HPK1 does not contain the small GTPase Rac1/Cdc42-binding domain and does not bind to either Rac1 or Cdc42, suggesting that HPK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HPK1 (Ser171)
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAP4K1
Gene Accession No. Q92918
Gene Alias EC 2.7.11.1; Hematopoietic progenitor kinase; hematopoietic progenitor kinase 1; HPK; HPK1; MAP4K1; MAPK/ERK kinase kinase kinase 1; MEK kinase kinase 1; MEKKK 1; mHPK1; mitogen activated protein kinase kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase kinase 1
Gene Symbols MAP4K1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S171 of human MAP4K1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11184
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.