Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-HSL (Ser554, Ser855) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538087
Description
In adipose tissue and heart, it primarily hydrolyzes stored triglycerides to free fatty acids, while in steroidogenic tissues, it principally converts cholesteryl esters to free cholesterol for steroid hormone production.
Specifications
Phospho-HSL (Ser554, Ser855) | |
Polyclonal | |
Unconjugated | |
LIPE | |
EC 3.1.1.79, Hormone sensitive lipase, LIPE, LIPS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16890, 25330, 3991 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q05469, P54310, P15304 | |
LIPE | |
A synthetic phosphopeptide derived from human HSL around the phosphorylation site of Ser855/554 (S-V-SP-E-A) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction