Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HSL (Ser563) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517488 Supplier Invitrogen™ Supplier No. PA517488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody. This antibody is not cross-reactive with Ser565 phosphorylated HSL.

Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HSL (Ser563)
Applications Western Blot
Classification Polyclonal
Concentration 22.48 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene LIPE
Gene Accession No. P54310
Gene Alias 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS
Gene Symbols LIPE
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pSer552 of human HSL (equivalent to pSer563 of rat HSL).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16890
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.