Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-HSL (Ser853) Recombinant Rabbit Monoclonal Antibody (6T6D8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535896
Description
Immunogen sequence: RRSVS.
Hormone-sensitive lipase (HSL) is an intracellular neutral lipase found in a variety of tissues, but primarily in adipose and steroidogenic tissues. It hydrolyzes triglycerides and cholesteryl esters. It is the major enzyme responsible for the mobilization of free fatty acids from adopise tissue.
Specifications
Phospho-HSL (Ser853) | |
Recombinant Monoclonal | |
0.08 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P54310 | |
LIPE | |
A synthetic phosphorylated peptide around S853 of human HSL (Q05469). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
6T6D8 | |
Unconjugated | |
LIPE | |
4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
16890 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction