Learn More
Invitrogen™ Phospho-HSL (Ser855, Ser554) Polyclonal Antibody

Description
Antibody detects endogenous levels of HSL only when phosphorylated at Ser855(Isoform 1) or Ser554(Isoform 2).
Specifications
Specifications
| Antigen | Phospho-HSL (Ser855, Ser554) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | LIPE |
| Gene Accession No. | P15304, P54310, Q05469 |
| Gene Alias | 4933403G17Rik; AOMS4; EC 3.1.1.79; FPLD6; hormone - sensitive lipase testicular isoform; hormone-sensitive lipase; hormone-sensitive lipase testicular isoform; HSL; HSL antibody; LHS; lipase E; lipase E, hormone sensitive type; lipase, hormone sensitive; lipase, hormone-sensitive; LIPE; LIPS |
| Gene Symbols | LIPE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.