Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HSP105 (Ser809) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564832 Supplier Invitrogen™ Supplier No. PA564832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-HSPH1 (Ser809) Polyclonal Antibody detects endogenous levels of HSPH1 only when phosphorylated at Ser809.

HSPH1 prevents the aggregation of denatured proteins in cells under severe stress, on which the ATP levels decrease markedly. This protein inhibits HSPA8/HSC70 ATPase and chaperone activities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HSP105 (Ser809)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSPH1
Gene Accession No. Q61699, Q66HA8, Q92598
Gene Alias 105kDa; 42 degrees C-HSP; AI790491; antigen NY-CO-25; heat shock 105/110 protein 1; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa; heat shock 105kDa/110kDa protein 1; heat shock 110 kDa protein; Heat shock protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein family H (Hsp110) member 1; heat shock protein, 105 kDa; heat shock-related 100 kDa protein E7I; heat-shock protein 110 kDa; HSP; Hsp105; HSP105 42 C-HSP; HSP105A; HSP105B; Hsp110; hsp110/105; HSP-E7I; Hsph1; I79_002075; Kiaa0201; KIAA0202; NY CO 25; NY-CO-25; SEP2; SEPT8; septin 8; SEPTIN8; septin-8
Gene Symbols HSPH1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSP105 (Phospho-Ser809).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10808, 15505, 288444
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.