Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-HTRA2 (Ser142) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564759 Supplier Invitrogen™ Supplier No. PA564759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-HTRA2 (Ser142) Polyclonal Antibody detects endogenous levels of HTRA2 only when phosphorylated at Ser142.

The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity (1-3). Recently, a serine protease has been isolated, which is released from the mitochondria upon induction of apoptosis by apoptotic stimuli. This protein called Omi/HtrA2 is a mitochondrial protein that binds to IAP (4). Like Smac/DIABLO, the mature Omi protein contains a conserved IAP-binding motif (AVPS) at its N terminus. The deregulated expression of Omi in the cytoplasm of mammalian cells induces apoptosis indicating that Omi could participate in the mitochondrial apoptotic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-HTRA2 (Ser142)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HTRA2
Gene Accession No. O43464, Q9JIY5
Gene Alias AI481710; High temperature requirement protein A2; HtrA serine peptidase 2; HtrA2; HtrA-like serine protease; mnd2; motor neuron degeneration 2; OMI; Omi stress-regulated endoprotease; PARK13; protease, serine, 25; Prss25; Serine protease 25; serine protease HTRA2, mitochondrial; serine protease OMI; serine proteinase OMI
Gene Symbols HTRA2
Host Species Rabbit
Immunogen A synthesized peptide derived from human HtrA2 (Phospho-Ser142).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27429, 297376, 64704
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.