Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-IGF1R (Tyr1346) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104773 Supplier Invitrogen™ Supplier No. PA5104773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of IGF1 Receptor only when phosphorylated at Tyrosine 1346.

IGF1R, also known as Insulin-like Growth Factor 1 (IGF-1) Receptor, is a receptor tyrosine kinase activated by IGF-1 and IGF-2. IGFR has been implicated in a variety of cancers. The protein is a tetramer consisting of two alpha, and two beta subunits. The alpha-subunits binds to insulin growth factor 1 with higher affinity than to insulin growth factor 2. Binding results in a conformational change, followed by autophosphorylation of IGF-1R at positions 1134, 1135, 1136. The activated IGF-1R in turn, causes phosphorylation of substrate proteins, followed by sequential activation of RAS, RAF, and mitogen-activated protein kinase isoforms ERK, p38, and JNK, leading to the transcription of genes that drive proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IGF1R (Tyr1346)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGF1R
Gene Accession No. P08069, P24062, Q60751
Gene Alias A330103N21Rik; CD221; CD221 antigen; D930020L01; hyft; IGF 1 receptor; IGF 1R; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols IGF1R
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGF1R(Accession P08069), corresponding to amino acid residues around phosphorylated Tyr1346.
Purification Method Sequential chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16001, 25718, 3480
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.