Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Phospho-IKK alpha (S176/S180) Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3768SP
This item is not returnable.
View return policy
Description
IKK alpha Monoclonal specifically detects IKK alpha in Human samples. It is validated for Western Blot.Specifications
IKK alpha (p ser180) | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O15111 | |
CHUK | |
Phosphopeptide containing the human IKK alpha S176/S180 Accession # O15111 | |
25 μg | |
Primary | |
Detects human IKK alpha when phosphorylated at S176/S180. | |
Human | |
Purified |
Western Blot | |
829177 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CHUK, conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IkBKalpha, IKK1, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKA, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1147 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction