Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ILK-1 (Ser246) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986570
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6570 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6570 Supplier Bioss Supplier No. BS5444R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen Phospho-ILK-1 (Ser246)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ILK
Gene Accession No. Q13418
Gene Alias 59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; Integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk
Gene Symbols ILK
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human ILK-1 around the phosphorylation site of Ser246.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.