Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phospho-ING5 (Thr152) Rat anti-Human, Unconjugated, Clone: 3H5, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABS130825UG
Description
phospho-ING5 (Thr152) Monoclonal antibody specifically detects phospho-ING5 (Thr152) in Human samples. It is validated for Western Blotting.
Specifications
phospho-ING5 (Thr152) | |
Monoclonal | |
Unconjugated | |
Inhibitor of growth protein 5;p28ING5 | |
Rat | |
∼32 kDa observed; 27.75 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
3H5 | |
NP_115705 | |
ING5 | |
A linear peptide surrounding phosphorylated Threonine 152 of human Inhibitor of growth protein 5 (ING5). | |
Protein G Purified | |
Signaling | |
Clone 3H5 is a rat monoclonal antibody that detects ING5 phosphorylated on Thr152 in human cells. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction