Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-IR/IGF1R (Tyr1165, Tyr1166, Tyr1189, Tyr1190) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110128 Supplier Invitrogen™ Supplier No. PA5110128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDYYRK.

Insulin and Insulin-like Growth Factor receptors are transmembrane tyrosine kinase receptors that play critical roles in development, cell growth and metabolism. Insulin/Insulin-like growth factor-1 binding to the extracellular domain of IR and IGFR leads to autophosphorylation of the receptor and activation of the intrinsic tyrosine kinase activity, which allows appropriate substrates to be phosphorylated. Phosphorylation sites that are unique to each receptor presumably play a key role in these signaling differences. The catalytic loops within the tyrosine kinase domains contain a three tyrosine motif corresponding to Tyr1158, 1162 and 1163 (for the IR) and Tyr1131, 1135 and 1136 (for the IGF1R). Autophosphorylation of the tyrosine sites within the activation loop proceeds in a processive manner initiating at the second tyrosine (1162 or 1135), followed by phosphorylation at tyrosines 1158 (IGF-1R tyrosine 1131), 1163 (IGF-1R tyrosine 1136), resulting in the full activation of IR and IGF1R.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-IR/IGF1R (Tyr1165, Tyr1166, Tyr1189, Tyr1190)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Insr
Gene Accession No. P06213, P08069, P15127, P15208, P24062, Q60751
Gene Alias 4932439J01Rik; A330103N21Rik; CD220; CD221; D630014A15Rik; D930020L01; HHF5; hyft; IGF1 Receptor; IGF-1 receptor; IGF1 Receptor beta; Igf1r; IGF-1R; IGF1R beta; IGF-I receptor; IGF-I Receptor beta; IGFIR; IGFIRC; IGFR; Igfr1; Insr; insulin like growth factor 1 receptor; insulin receptor; insulin receptor precursor; insulin receptor preproprotein; Insulin receptor subunit alpha; Insulin receptor subunit beta; Insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor receptor type 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; IR; IR-A; IR-B; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Gene Symbols IGF1R, Insr
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around Y1165 & Y1166 of human IGF1RIGF1R (NP_0008661).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16001, 16337, 24954, 25718, 3480, 3643
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.