Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IRAK1 (Thr209) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985210
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5210 Supplier Bioss Supplier No. BS3193R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interleukin-1 receptor-associated kinase 1 (IRAK1) is a serine/threonine-protein kinase that plays a critical role in initiating innate immune response against foreign pathogens. IRAK is associated with Toll-like receptor (TLR) and IL-1R signaling pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK′s association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.

Specifications

Antigen Phospho-IRAK1 (Thr209)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Irak1
Gene Accession No. P51617
Gene Alias AA408924; Il1rak; interleukin 1 receptor associated kinase 1; Interleukin-1 receptor-associated kinase 1; interleukin-1 receptor-associated kinase-1; IRAK; Irak1; IRAK-1; IRAK1-S; mPLK; pelle; Pelle homolog; Pelle-like protein kinase; Plpk; RGD1563841
Gene Symbols Irak1
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human IRAK1 around the phosphorylation site of Thr209 [RG (p-T)HN].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3654
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.