Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IRAK4 (Thr345) Monoclonal Antibody (A8A8), Abbomax

Catalog No. 502203907
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3907 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3907 Supplier Abbomax Inc Supplier No. 600560
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Jurkat Cell lysate Cellular location: Cytoplasmic.

Phospho-IRAK4 (Thr345) Monoclonal antibody specifically detects Phospho-IRAK4 (Thr345) in Bovine, Canine, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Phospho-IRAK4 (Thr345)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone A8A8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene IRAK4
Gene Accession No. Q1RMT8, Q8R4K2, Q9NWZ3
Gene Alias 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64
Host Species Mouse
Immunogen A synthetic peptide surrounding to the epitope -VMTSR- with a phosphorylation site at Thr345 of human IRAK-4 protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266632, 486601, 51135, 533692
Target Species Bovine, Canine, Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.