Learn More
Invitrogen™ Phospho-IRF3 (Ser386) Polyclonal Antibody

Description
Positive Control: HeLa, HeLa treat with CIP Predicted Reactivity: Rhesus Monkey (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Phospho-IRF3 (Ser386) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | IRF3 |
| Gene Accession No. | Q14653 |
| Gene Alias | C920001K05Rik; IIAE7; Interferon regulatory factor 3; IRF3; IRF-3; IRF3 (phospho S386); IRF3 (pS386); IRF3 (pSer386); IRF3 phospho; IRF3 phospho S386; IRF3 phospho Ser386; phospho IRF3 |
| Gene Symbols | IRF3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.