Learn More
Invitrogen™ Phospho-IRS1 (Ser794) Polyclonal Antibody

Description
This antibody is predicted to react with human and mouse based on sequence homology.
Specifications
Specifications
| Antigen | Phospho-IRS1 (Ser794) |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | IRS1 |
| Gene Accession No. | P35568, P35569, P35570 |
| Gene Alias | G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 |
| Gene Symbols | IRS1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.