Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ phospho-IRS1 (Tyr941), Rabbit, Unlabeled, Polyclonal,

Catalog No. 07848I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
07848I 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07848I Supplier MilliporeSigma™ Supplier No. 07848I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Insulin receptor substrate 1 (UniProt P35568; also known as IRS-1) is encoded by the IRS1 gene (Gene ID 3667) in human. The insulin receptor substrates (IRS-1, IRS-2, IRS-3, and IRS-4) are adaptor proteins involved in modulating cell growth, metabolism, survival, and differentiation by transducing receptor signaling to multiple downstream effectors. IRS family proteins contain a conserved N-terminal pleckstrin homology (PH) domain (a.a. 12-115 of human IRS-1), a phosphotyrosine-binding (PTB) domain (a.a. 160-264 of human IRS-1), and multiple tyrosine phosphorylation sites in the C-terminal region, including Tyr612, Tyr896, Tyr941, Tyr1158, and Tyr1220. The PTB domain meidates the interaction of IRS-1, -2 and -3 with IR juxtamembrane NPXYp motif, facilitating subsequent receptor-mediated IRS tyrosine phosphorylation. Upon phosphorylation, IRS proteins serve as binding sites for Src homology 2 (SH2) domain-containing proteins, such as SHP-2 and PI 3-kinase. Functions of IRS proteins are largely dependent on the phosphorylations of specific tyrosine residues, which in turn are subject to negative regulation by phosphatase activity. In addition, IRS proteins can be phosphorylated by serine/threonine kinases, such as PI-3 kinase, Akt/PKB, GSK-3, mTOR, and PKC, and increased Ser/Thr phosphorylation reduces their binding to IR and tyrosine phosphorylation. Inhibition of PI 3-kinase enhances insulin-stimulated IRS-1 Tyr612/Tyr941 phosphorylation (p85-binding sites) and increases IRS-1 association with PI 3-kinase p85 subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-IRS1 (Tyr941)
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene IRS1
Gene Accession No. P35568
Host Species Rabbit
Immunogen KLH-conjugated phosphopeptide corresponding to human IRS1 target region sequence containing phosphorylated Tyr941.
Purification Method Affinity Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005535
Test Specificity This polyclonal antibody detected a ∽165 kDa target band in lysates from serum-starved HEK293 cells only after, but not before, insulin stimulation. Target region sequence is not 100% conserved among human, mouse, and rat species.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.