Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ITCH / AIP4 (Tyr420) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5105078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5105078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5105078 Supplier Thermo Scientific Supplier No. PA5105078
Add to Cart
Add to Cart

Phospho-ITCH / AIP4 (Tyr420) Polyclonal Antibody

Phospho-ITCH / AIP4 (Tyr420) Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-ITCH / AIP4 (Tyr420)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ITCH
Gene Accession No. Q8C863, Q96J02
Gene Alias 6720481N21Rik; 8030492O04Rik; A130065M08; ADMFD; AIF4; AIP4; atrophin-1 interacting protein 4; Atrophin-1-interacting protein 4; C230047C07Rik; dJ468O1.1; E3 ubiquitin-protein ligase Itchy; E3 ubiquitin-protein ligase Itchy homolog; HECT-type E3 ubiquitin transferase Itchy homolog; ITCH; itch E3 ubiquitin ligase; itchy E3 ubiquitin protein ligase; itchy E3 ubiquitin protein ligase homolog; itchy, E3 ubiquitin protein ligase; NAPP1; NFE2-associated polypeptide 1
Gene Symbols ITCH
Host Species Rabbit
Immunogen A synthesized peptide derived from human ITCH(Accession Q96J02), corresponding to amino acid residues around phosphorylated Tyr420.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16396, 83737
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.