Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ITGB3 (Tyr785) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986575
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6575 Supplier Bioss Supplier No. BS5452R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD51 and CD61 molecules (alpha V and beta 3 integrins) form a complex. This complex binds vitronection at the RGD sequence and can also bind fibrinogen, von Willebrand factor, thrombospondin, fibronectin, osteopontin and collagen. Reacts with osteoclasts, placenta, melanoma cell lines and weakly with platelets.

Specifications

Antigen Phospho-ITGB3 (Tyr785)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ITGB3
Gene Accession No. P05106, P06756
Gene Alias antigen CD61; antigen identified by monoclonal antibody L230; BDPLT16; BDPLT2; CD51; CD61; Dnmt3l-ps1 pseudogene; GP3A; GPIIIa; GT; integrin; integrin alpha V; Integrin alpha-V; Integrin alpha-V heavy chain; Integrin alpha-V light chain; integrin alphaVbeta3; integrin beta 3; integrin beta 3 (Cd61); Integrin beta-3; integrin subunit alpha V; integrin subunit beta 3; integrin, alpha V; integrin, alpha V (vitronectin receptor, alpha polypeptide, antigen CD51); integrin, beta 3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); ITGAV; Itgb3; MSK8; Platelet membrane glycoprotein IIIa; Vitronectin receptor; vitronectin receptor alpha subunit; Vitronectin receptor subunit alpha; VNRA; VTNR
Gene Symbols ITGAV, ITGB3
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human ITGB3 around the phosphorylation site of Tyr785.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685, 3690
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.