Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-JAK1 (Tyr1034, Tyr1035) Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc PJAK1140AP

Description
Phospho-JAK1 (Tyr1034, Tyr1035) Polyclonal antibody specifically detects Phospho-JAK1 (Tyr1034, Tyr1035) in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Phospho-JAK1 (Tyr1034, Tyr1035) | |
Polyclonal | |
Unconjugated | |
JAK1 | |
AA960307; BAP004; C130039L05Rik; EC 2.7.10.2; Jak1; JAK-1; JAK1A; JAK1B; Janus kinase 1; Janus protein tyrosine kinase 1; JTK3; kinase Jak1; Tyrosine-protein kinase JAK1 | |
Synthetic peptide taken within amino acid region 1000-1050 on human JAK1 protein. Phosphorylated 1034 & 1035 tyrosine residues. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P23458, P52332 | |
Rabbit | |
Affinity chromatography | |
RUO | |
16451, 3716, 84598 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction