Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-JAK2/JAK3 (Tyr966, Tyr939) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567508 Supplier Invitrogen™ Supplier No. PA567508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Rat samples.

Janus Activating Kinase 2 and 3 (JAK2/JAK3) is one of a number of related tyrosine kinases involved in cytoplasmic signal transduction. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-JAK2/JAK3 (Tyr966, Tyr939)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene JAK2
Gene Accession No. O60674, P52333, Q62120, Q62137, Q62689, Q63272
Gene Alias Breast-JAK3; fae; Fd17; JAK2; JAK-2; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; JTK10; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; THCYT3; tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3; wil
Gene Symbols JAK2, JAK3
Host Species Rabbit
Immunogen Synthesized peptide derived from human JAK2/JAK3 around the phosphorylation site of Tyr966/939.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16452, 16453, 24514, 25326, 3717, 3718
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.