Learn More
Invitrogen™ Phospho-Jak2 (Tyr1007, Tyr1008) Recombinant Rabbit Monoclonal Antibody (JAK2Y10071008-PB6), PE, Invitrogen™
Description
Specifications
Specifications
| Antigen | Phospho-Jak2 (Tyr1007, Tyr1008) |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | JAK2Y10071008-PB6 |
| Conjugate | PE |
| Formulation | PBS with 0.20% BSA and 0.09% sodium azide |
| Gene | JAK2 |
| Gene Accession No. | O60674 |
| Gene Alias | EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 |
| Gene Symbols | JAK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.