Learn More
Invitrogen™ Phospho-JAK2 (Tyr1008) Polyclonal Antibody

Description
Immunogen sequence: EYYKV; Positive Samples: Jurkat; Cellular Location: Cytoplasm, Endomembrane system, Nucleus, Peripheral membrane protein.
Specifications
Specifications
| Antigen | Phospho-JAK2 (Tyr1008) |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | JAK2 |
| Gene Accession No. | O60674 |
| Gene Alias | EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 |
| Gene Symbols | JAK2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.