Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JMJD2B (Thr305) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982032
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2032 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2032 Supplier Bioss Supplier No. BS12200R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.

Specifications

Antigen Phospho-JMJD2B (Thr305)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KDM4B
Gene Accession No. O94953
Gene Alias 4732474L06Rik; JHDM3B; JmjC domain-containing histone demethylation protein 3B; Jmjd2b; jumonji domain containing 2B; Jumonji domain-containing protein 2B; KDM4B; KIAA0876; lysine (K)-specific demethylase 4B; lysine demethylase 4B; Lysine-specific demethylase 4B; mKIAA0876; TDRD14B; tudor domain containing 14B
Gene Symbols KDM4B
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human JMJD2B around the phosphorylation site of Thr305.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23030
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.