Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Phospho-JNK1/JNK2 (Thr183, Tyr185) Monoclonal Antibody (F.971.6)
GREENER_CHOICE

Catalog No. PIMA514943
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514943 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514943 Supplier Invitrogen™ Supplier No. MA514943
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-JNK1/JNK2 (Thr183, Tyr185)
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F.971.6
Concentration 293 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5
Gene MAPK9
Gene Accession No. P45983, P45984, P49185, P49186, Q91Y86, Q9WTU6
Gene Alias AI849689; AI851083; C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; Mapk8; Mapk9; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; OTTHUMP00000019552; p54 gamma; p54a; p54-alpha; p54aSAPK; Phospho-JNK; PRKM; PRKM8; Prkm9; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK; SAPK 1; SAPK gamma; SAPK1; SAPK1A; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1c; stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2
Gene Symbols MAPK8, MAPK9
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pThr183/Tyr185 of human SAPK/JNK.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116554, 26419, 26420, 50658, 5599, 5601
Target Species Drosophila, Human, Mouse, Rat, Yeast
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.