Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JunB (Ser259) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986584
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6584 Supplier Bioss Supplier No. BS5464R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JunB ,a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5′-TGA[CG]TCA-3′. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.

Specifications

Antigen Phospho-JunB (Ser259)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Junb
Gene Accession No. P17275
Gene Alias activator protein 1; AP-1; Jun B; jun B proto-oncogene; JUNB; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; MyD21; Oncogene Jun-B; Transcription factor jun-B
Gene Symbols Junb
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human JunB around the phosphorylation site of Ser259 [PV (p-S)PI].
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3726
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.