Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JunD (Ser255) Polyclonal Antibody, Abbomax

Catalog No. 502203571
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3571 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3571 Supplier Abbomax Inc Supplier No. 50011784
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human mammary gland tissue and lung squamous cell carcinoma tissue; Cellular location: Nucleus.

Phospho-JunD (Ser255) Polyclonal antibody specifically detects Phospho-JunD (Ser255) in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Specifications

Antigen Phospho-JunD (Ser255)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 25mM tris with proprietary stabilizer and 0.01% sodium azide
Gene JUND
Gene Accession No. P15066, P17535
Gene Alias activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D
Host Species Rabbit
Immunogen A synthetic peptide of human JunD with a phosphrylation site Serine 255
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16478, 3727
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.