Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-JunD (Ser255) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104821
Description
Antibody detects endogenous levels of JunD only when phosphorylated at Serine 255.
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
Specifications
Phospho-JunD (Ser255) | |
Polyclonal | |
Unconjugated | |
JUND | |
activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
Rabbit | |
sequential chromatography | |
RUO | |
16478, 24518, 3727 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P15066, P17535, P52909 | |
JUND | |
A synthesized peptide derived from human JUND(Accession P17535), corresponding to amino acid residues around phosphorylated Ser255. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction