Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Phospho-JunD (Ser255) Recombinant Rabbit Monoclonal Antibody (5M8M4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551532
Description
Immunogen sequence: GESPP.
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
Specifications
Phospho-JunD (Ser255) | |
Recombinant Monoclonal | |
0.52 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P17535 | |
JUND | |
A synthetic phosphorylated peptide around S255 of human Phospho-JunD-S255 (NP_005345.3). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Western Blot | |
5M8M4 | |
Unconjugated | |
JUND | |
activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
Rabbit | |
Affinity chromatography | |
RUO | |
3727 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction