Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JUP (Tyr550) Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986586
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6586 Supplier Bioss Supplier No. BS5467R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.

Specifications

Antigen Phospho-JUP (Tyr550)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene JUP
Gene Accession No. P14923
Gene Alias ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; CTNNG; D930025P04Rik; Desmoplakin III; Desmoplakin-3; DP3; DPIII; gamma-catenin; gamma-catenin (plakoglobin); Junction plakoglobin; Jup; OTTHUMP00000164735; PDGB; PG; PKGB
Gene Symbols JUP
Host Species Rabbit
Immunogen KLH conjugated synthetic phosphopeptide derived from human JUP around the phosphorylation site of Tyr550.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3728
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.