Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-KIRREL (Tyr737, Tyr738) Rabbit anti-Chicken, Human, Mouse, Rat, Zebrafish, Polyclonal, FabGennix

Catalog No. 502208615
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8615 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8615 Supplier Fabgennix Inc Supplier No. PNEPH140AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phospho-KIRREL (Tyr737, Tyr738) Polyclonal antibody specifically detects Phospho-KIRREL (Tyr737, Tyr738) in Chicken, Human, Mouse, Rat, Zebrafish samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Phospho-KIRREL (Tyr737, Tyr738)
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene KIRREL1
Gene Accession No. Q6X936, Q80W68, Q96J84
Gene Alias 6720469N11Rik; FLJ10845; kin of IRRE like (Drosophila); kin of IRRE like 1; Kin of irregular chiasm-like protein 1; kin of IRRE-like protein 1; Kirrel; KIRREL1; MGC129542; MGC129543; NEPH1; nephrin 1; nephrin related; nephrin-like protein 1
Host Species Rabbit
Immunogen Phosphorylated synthetic peptide with phospo tyrosine 737 and 738 on mouse Kin of IRRE-like protein 1.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170643, 310695, 425302, 55243
Target Species Chicken, Human, Mouse, Rat, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.